Search
Search
| Gene Symbol: Orc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011959.2 | NP_036089.1 | 4 - 5 | 512 | 117 | |
| GenBank mRNA | AJ007360.1 | - | 4 - 5 | 509 | 117 | |
| AK084683.1 | - | 4 - 5 | 540 | 117 | ||
| AK144983.1 | - | 4 - 5 | 528 | 117 | ||
| AK165912.1 | - | 4 - 5 | 518 | 117 | ||
| AK166537.1 | - | 4 - 5 | 451 | 117 | ||
| AK169683.1 | - | 4 - 5 | 486 | 117 | ||
| BC006927.1 | - | 4 - 5 | 528 | 3601908 | 117 | |
Set Membership: |
|