Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01167756_s1
          See other HIST1H3D GE Assays ›
          Gene Symbol:
          Hist1h3d
          Gene Name:
          histone cluster 1, H3d
          Gene Aliases:
          H3-b
          Chromosome Location:
          Chr.13: 23575763 - 23576266 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01167756_s1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hist1h3d
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK020421.1 - 1168 77

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 71 and 85 Mus musculus 1700 Inventoried

          Panther Classification:

          Molecular Function -

          histone

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28882318 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2127139 NM_178204.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2127139 NM_178204.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28882315 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28882317 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28882314 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28882313 - -
          Whole Mouse Genome G4122A A_51_P492488 NM_178204.1 mouse_transcriptome
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10589471 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28882318 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882313 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882305 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882342 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882359 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882254 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882293 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882338 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882350 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882257 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882351 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882249 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882306 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882357 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882275 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882362 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882251 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882282 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882297 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882259 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882242 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882246 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882252 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882291 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882348 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882295 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882315 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882317 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882255 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882354 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882352 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882340 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882289 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882240 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882253 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882344 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882337 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28882315 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28882317 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28882313 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28882312 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10404066 - -
          Affymetrix Mouse Gene 1.0 ST Array 10408248 - -
          Affymetrix Mouse Gene 1.0 ST Array 10404029 - -
          Affymetrix Mouse Gene 1.0 ST Array 10404043 - -
          Affymetrix Mouse Gene 1.0 ST Array 10408203 - -
          Affymetrix Mouse Gene 1.0 ST Array 10408084 - -
          Affymetrix Mouse Gene 1.0 ST Array 10494400 - -
          Affymetrix Mouse Gene 1.0 ST Array 10494406 - -
          Affymetrix Mouse Gene 1.0 ST Array 10494403 - -
          Affymetrix Mouse Gene 1.0 ST Array 10408240 - -
          Affymetrix Mouse Gene 1.0 ST Array 10403942 - -
          Affymetrix Mouse Gene 1.0 ST Array 10404050 - -
          Affymetrix HT MG-430 PM Array Plate 1453573_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10408083 - -
          Affymetrix Mouse Gene 1.1 ST Array 10408239 - -
          Affymetrix Mouse Gene 1.1 ST Array 10403941 - -
          Affymetrix Mouse Gene 1.1 ST Array 10404028 - -
          Affymetrix Mouse Gene 1.1 ST Array 10408246 - -
          Affymetrix Mouse Gene 1.1 ST Array 10408202 - -
          Affymetrix Mouse Gene 1.1 ST Array 10404038 - -
          Affymetrix Mouse Gene 1.1 ST Array 10404049 - -
          Affymetrix Mouse Gene 1.1 ST Array 10404065 - -
          Mouse Genome 430 2.0 Array 1453573_at - -
          Mouse Genome 430 2.0 Array 1453573_PM_at - -
          Mouse Genome 430 2.0 Array 1427863_PM_at - -
          Mouse Genome 430 2.0 Array 1427863_at - -
          Mouse Genome 430 2.0 Array 1427864_PM_at - -
          Mouse Genome 430 2.0 Array 1427864_at - -
          Affymetrix Mouse Clariom S Array PSR1300001370.mm.1 NM_178204.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1300001370.mm.1 TC1300000240.mm.1 MTA TC
          Mouse Expression Set 430 1453573_at - -
          Mouse Expression Set 430 1418366_at - -
          Mouse Expression Set 430 1427864_PM_at - -
          Mouse Expression Set 430 1427864_at - -
          Mouse Expression Set 430 1460314_s_at - -
          Mouse Expression Set 430 1418366_PM_at - -
          Mouse Expression Set 430 1418367_PM_x_at - -
          Mouse Expression Set 430 1460314_PM_s_at - -
          Mouse Expression Set 430 1418367_x_at - -
          Mouse Expression Set 430 1453573_PM_at - -
          Mouse Expression Set 430 1442051_at - -
          Mouse Expression Set 430 1442051_PM_at - -
          Murine Genome U74v2 Set 96416_f_at - -
          Murine Genome U74v2 Set 93067_f_at - -
          Murine Genome U74v2 Set 93068_r_at - -
          Murine Genome U74v2 Set 94756_at - -
          Murine Genome U74v2 Set 96416_PM_f_at - -
          Murine Genome U74v2 Set 93022_PM_i_at - -
          Murine Genome U74v2 Set 93023_f_at - -
          Murine Genome U74v2 Set 93022_i_at - -
          Murine Genome U74v2 Set 94756_PM_at - -
          Murine Genome U74v2 Set 93023_PM_f_at - -
          Murine Genome U74v2 Set 93067_PM_f_at - -
          Murine Genome U74v2 Set 93068_PM_r_at - -
          Mouse Genome 430A 2.0 Array 1427864_PM_at - -
          Mouse Genome 430A 2.0 Array 1460314_s_at - -
          Mouse Genome 430A 2.0 Array 1418367_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1418367_x_at - -
          Mouse Genome 430A 2.0 Array 1418366_PM_at - -
          Mouse Genome 430A 2.0 Array 1460314_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1427864_at - -
          Mouse Genome 430A 2.0 Array 1418366_at - -
          Mouse Genome 430A 2.0 Array 1453573_PM_at - -
          Mouse Genome 430A 2.0 Array 1453573_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 388844 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 70687 - -
          Illumina mouse-6 v1.0 expression beadchip 103130494 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000070687 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2729958 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1231671 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2729958 - -
          mouse-6 v1.1 (Illumina) scl0319149.1_41-S - -
          mouse-6 v1.1 (Illumina) scl00319149.2_275-S - -
          Illumina Sentrix mouse-6.v1 bead array 70687 - -
          Illumina Sentrix mouse-6.v1 bead array 103130494 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl00319149.2_275-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 70687 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 103130494 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2729958 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1231671 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2729958 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl00319149.2_275-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline