Search
Search

Egfl6| Gene Symbol: Egfl6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019397.3 | NP_062270.1 | 3 - 4 | 465 | 146 | |
| GenBank mRNA | AJ245672.1 | - | 3 - 4 | 467 | 146 | |
| AK053738.1 | - | 3 - 4 | 462 | 146 | ||
| AK078534.1 | - | 3 - 4 | 443 | 146 | ||
| BC117702.2 | - | 3 - 4 | 451 | 40062530 | 146 | |
Set Membership: |
|