Search
Search
| Gene Symbol: Zp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011775.6 | NP_035905.1 | 4 - 5 | 348 | 75 | |
| GenBank mRNA | AK135903.1 | - | 4 - 5 | 348 | 75 | |
| AK138699.1 | - | 4 - 5 | 349 | 75 | ||
| BC071183.1 | - | 4 - 5 | 330 | 30473497 | 75 | |
| M34148.1 | - | 4 - 5 | 348 | 75 | ||
Set Membership: |
|