Search
Search

Kdm7a| Gene Symbol: Kdm7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033430.4 | NP_001028602.2 | 1 - 2 | 331 | 76 | |
| GenBank mRNA | AK042327.1 | - | 1 - 2 | 348 | 76 | |
| AK042834.1 | - | 1 - 2 | 332 | 76 | ||
| AK136209.1 | - | 1 - 2 | 170 | 76 | ||
| AK136238.1 | - | 1 - 2 | 318 | 76 | ||
| BC145216.1 | - | 1 - 2 | 221 | 76 | ||
| BC145848.1 | - | 1 - 2 | 221 | 40131151 | 76 | |
Set Membership: |
|