Search
Search

Tbxas1| Gene Symbol: Tbxas1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011539.3 | NP_035669.3 | 10 - 11 | 1390 | 97 | |
| GenBank mRNA | AK139479.1 | - | 10 - 11 | 1353 | 97 | |
| BC029014.1 | - | 10 - 11 | 1392 | 5007629 | 97 | |
| L18868.1 | - | 10 - 11 | 1261 | 97 | ||
Set Membership: |
|