Search
Search
| Gene Symbol: Upk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC104260.1 | - | 3 - 4 | 405 | 40039813 | 76 |
| BC104261.1 | - | 3 - 4 | 405 | 40039814 | 76 | |
| DQ358978.1 | - | 3 - 4 | 344 | 76 | ||
| U08030.1 | - | 3 - 4 | 403 | 76 | ||
Set Membership: |
|