Search
Search

Nlrp5| Gene Symbol: Nlrp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF074018.1 | - | 2 - 3 | 157 | 154 | |
| AK087774.1 | - | 3 - 4 | 225 | 154 | ||
| AY196361.1 | - | 2 - 3 | 157 | 154 | ||
| AY196362.1 | - | 2 - 3 | 157 | 154 | ||
| AY329484.1 | - | 2 - 3 | 157 | 154 | ||
| AY329485.1 | - | 2 - 3 | 157 | 154 | ||
| AY329486.1 | - | 2 - 3 | 157 | 154 | ||
| AY329487.1 | - | 2 - 3 | 157 | 154 | ||
| AY329488.1 | - | 2 - 3 | 157 | 154 | ||
| AY329489.1 | - | 2 - 3 | 157 | 154 | ||
| AY329490.1 | - | 2 - 3 | 157 | 154 | ||
| AY329491.1 | - | 2 - 3 | 157 | 154 | ||
| BC053384.1 | - | 3 - 4 | 224 | 30049556 | 154 | |
Set Membership: |
|