Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Mm01168771_m1
See other MCU GE Assays ›
Gene Symbol:
Mcu
Gene Name:
mitochondrial calcium uniporter
Gene Aliases:
2010012O16Rik, AV064928, C10orf42, Ccdc109a, D130073L02Rik, Gm64
Chromosome Location:
Chr.10: 59446984 - 59616692 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID
Mm01168771_m1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2,900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2,900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2,900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: Mcu
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001033259.4
NP_001028431.2
1 - 2
169
96
GenBank mRNA
AK144727.1
-
1 - 2
170
96
Back To Top
More Information
Updated Mapping Information
This assay targets the same location as MTA transcript cluster(s) TC1000002212.mm.1
Set Membership:
Amplicon length between 86 and 100
Mus musculus
Probe spans exons
Gene Ontology Categories:
Function(s)
Process(es)
transport
ion transport
calcium ion transport
mitochondrial calcium ion transport
calcium-mediated signaling
positive regulation of insulin secretion
protein complex oligomerization
calcium ion transmembrane import into mitochondrion
glucose homeostasis
mitochondrial calcium ion homeostasis
positive regulation of mitochondrial calcium ion concentration
calcium ion transmembrane transport
calcium channel activity
protein binding
uniporter activity
identical protein binding
MicroArray Probes:
Affymetrix
Array Name
Probe IDs
Targets
Target Type
Affymetrix Mouse Clariom S Array
PSR1000015516.mm.1
NM_001033259.4.1
Refseq
Affymetrix Mouse Clariom S Array
PSR1000015516.mm.1
NM_001033259.4
Refseq
Affymetrix Mouse Clariom S Array
PSR1000015516.mm.1
TC1000002212.mm.1
MTA TC
Affymetrix Mouse Clariom S Array
PSR1000015509.mm.1
NM_001033259.4.1
Refseq
Affymetrix Mouse Clariom S Array
PSR1000015509.mm.1
NM_001033259.4
Refseq
Affymetrix Mouse Clariom S Array
PSR1000015509.mm.1
TC1000002212.mm.1
MTA TC
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-744bc48644-nrcn4:80/100.66.78.247:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline