Search
Search
| Gene Symbol: Bcr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081412.2 | NP_001074881.1 | 14 - 15 | 2909 | 130 | |
| GenBank mRNA | AK129482.1 | - | 11 - 12 | 1099 | 130 | |
| BC048842.1 | - | 2 - 3 | 153 | 130 | ||
| BC060270.1 | - | 14 - 15 | 2147 | 130 | ||
| BC131682.1 | - | 15 - 16 | 2314 | 40111128 | 130 | |
Set Membership: |
|