Search
Search

Rufy2| Gene Symbol: Rufy2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027425.3 | NP_081701.2 | 2 - 3 | 289 | 100 | |
| GenBank mRNA | AK045485.1 | - | 2 - 3 | 290 | 100 | |
| AK083205.1 | - | 2 - 3 | 274 | 100 | ||
| AK173195.1 | - | 2 - 3 | 250 | 100 | ||
| BB643897.1 | - | 2 - 3 | 290 | 100 | ||
| BC017645.1 | - | 2 - 3 | 251 | 100 | ||
| BC038265.1 | - | 2 - 3 | 230 | 100 | ||
| BC137736.1 | - | 2 - 3 | 238 | 8860755 | 100 | |
| BC137737.1 | - | 2 - 3 | 238 | 8860756 | 100 | |
Set Membership: |
|