Search
Search

Tbx18| Gene Symbol: Tbx18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023814.4 | NP_076303.3 | 2 - 3 | 934 | 57 | |
| GenBank mRNA | AF306666.1 | - | 2 - 3 | 869 | 57 | |
| AK032660.1 | - | 2 - 3 | 936 | 57 | ||
| BC145691.1 | - | 2 - 3 | 831 | 40130994 | 57 | |
Set Membership: |
|