Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01169164_g1
          See other MMP1B GE Assays ›
          Gene Symbol:
          Mmp1b
          Gene Name:
          matrix metallopeptidase 1b (interstitial collagenase)
          Gene Aliases:
          Mcol-B, Mcolb
          Chromosome Location:
          Chr.9: 7367640 - 7388026 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01169164_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mmp1b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_032007.3 NP_114396.3 9 - 10 1322 125
          GenBank mRNA AJ278461.1 - 9 - 10 1291 125

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900001718.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Protease 3' Most Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mmp1b

          Panther Classification:

          Molecular Function -

          metalloprotease

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          collagen catabolic process
          metalloendopeptidase activity
          calcium ion binding
          peptidase activity
          metallopeptidase activity
          zinc ion binding
          hydrolase activity
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2105643 NM_032007.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 297276 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 22597 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 444759 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 928284 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 613810 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 845136 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 667067 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 128617 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 159872 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 88085 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P127689 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P145423 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2105643 NM_032007.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26287905 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26287876 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26287882 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26287891 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26287892 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26287879 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26287889 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06692975 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P145423 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P127689 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P134006 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P145423 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P127689 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07712281 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08196984 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07712282 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07712284 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26287866 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26287864 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26287806 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26287871 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26287877 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26287879 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26287798 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26287813 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26287805 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26287801 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26287860 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26287833 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26287804 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26287841 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26287869 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26287838 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26287831 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26287835 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26287840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26287865 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26287808 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26287870 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26287809 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26287845 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26287821 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26287891 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26287793 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26287790 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26287826 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26287873 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26287847 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26287789 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26287867 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26287794 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26287848 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26287819 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26287817 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26287843 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26287859 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26287791 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26287799 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26287862 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07938434 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07712282 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26287866 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26287887 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26287871 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26287859 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26287870 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26287905 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26287891 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26287869 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26287882 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26287892 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26287895 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26287873 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26287867 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26287864 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26287876 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26287877 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26287865 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10590789 - -
          Affymetrix Mouse Gene 1.0 ST Array 10590788 - -
          Affymetrix Mouse Gene 1.0 ST Array 10590787 - -
          Affymetrix Mouse Gene 1.0 ST Array 10590785 - -
          Affymetrix Mouse Gene 1.0 ST Array 10590783 - -
          Affymetrix Mouse Gene 1.0 ST Array 10590782 - -
          Affymetrix Mouse Gene 1.0 ST Array 10590790 - -
          Affymetrix Mouse Gene 1.0 ST Array 10590784 - -
          Affymetrix Mouse Gene 1.0 ST Array 10590786 - -
          Affymetrix HT MG-430 PM Array Plate 1422273_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10590781 - -
          Mouse Genome 430 2.0 Array 1422273_at - -
          Mouse Genome 430 2.0 Array 1422273_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 83996_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 83996_at - -
          Affymetrix Mouse Clariom S Array PSR0900014495.mm.1 NM_032007.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900014495.mm.1 NM_032007.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900014495.mm.1 TC0900001718.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900014498.mm.1 NM_032007.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900014498.mm.1 NM_032007.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900014498.mm.1 TC0900001718.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900014499.mm.1 NM_032007.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900014499.mm.1 NM_032007.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900014499.mm.1 TC0900001718.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900014497.mm.1 NM_032007.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900014497.mm.1 NM_032007.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900014497.mm.1 TC0900001718.mm.1 MTA TC
          Mouse Expression Set 430 1422273_at - -
          Mouse Expression Set 430 1422273_PM_at - -
          Mouse Genome 430A 2.0 Array 1422273_at - -
          Mouse Genome 430A 2.0 Array 1422273_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1422273_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1422273_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 780722 - -
          Illumina mouse-6 v1.0 expression beadchip 780722 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000780722 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1236455 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1236455 - -
          mouse-6 v1.1 (Illumina) scl36263.10.1_19-S - -
          Illumina Sentrix mouse-6.v1 bead array 780722 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl36263.10.1_19-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 780722 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1236455 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1236455 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline