Search
Search

Osbpl11| Gene Symbol: Osbpl11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_176840.3 | NP_789810.2 | 5 - 6 | 1422 | 66 | |
| GenBank mRNA | BC035278.1 | - | 5 - 6 | 1422 | 5043236 | 66 |
Set Membership: |
|