Search
Search

Grin3b| Gene Symbol: Grin3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130455.2 | NP_569722.1 | 1 - 2 | 604 | 115 | |
| GenBank mRNA | AF396649.1 | - | 1 - 2 | 613 | 115 | |
| AY062435.1 | - | 1 - 2 | 604 | 115 | ||
Set Membership: |
|