Search
Search
| Gene Symbol: Sik2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178710.3 | NP_848825.2 | 10 - 11 | 1686 | 122 | |
| GenBank mRNA | AB067780.1 | - | 10 - 11 | 1587 | 122 | |
| AK090386.1 | - | 10 - 11 | 1587 | 122 | ||
Set Membership: |
|