Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Mapk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038663.1 | NP_001033752.1 | 4 - 5 | 846 | 75 | |
NM_011949.3 | NP_036079.1 | 4 - 5 | 846 | 75 | ||
GenBank mRNA | AK035386.1 | - | 4 - 5 | 820 | 75 | |
AK048127.1 | - | 4 - 5 | 848 | 75 | ||
AK087925.1 | - | 4 - 5 | 821 | 75 | ||
AK132241.1 | - | 4 - 5 | 834 | 75 | ||
AK148853.1 | - | 4 - 5 | 826 | 75 | ||
AK193200.1 | - | 1 - 2 | 37 | 75 | ||
BC058258.1 | - | 4 - 5 | 789 | 6468233 | 75 | |
D10939.1 | - | 4 - 5 | 661 | 75 | ||
X58712.1 | - | 4 - 5 | 619 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |