Search
Search

Aifm3| Gene Symbol: Aifm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291070.1 | NP_001277999.1 | 15 - 16 | 1630 | 68 | |
| NM_175178.4 | NP_780387.2 | 17 - 18 | 2036 | 68 | ||
| GenBank mRNA | AK158809.1 | - | 16 - 17 | 1698 | 68 | |
| BC046517.1 | - | 17 - 18 | 2023 | 68 | ||
| BC096476.1 | - | 16 - 17 | 1811 | 4221224 | 68 | |
Set Membership: |
|