Search
Search

Numbl| Gene Symbol: Numbl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010950.2 | NP_035080.2 | 6 - 7 | 718 | 102 | |
| GenBank mRNA | AK147523.1 | - | 4 - 5 | 6505 | 102 | |
| BC059915.1 | - | 5 - 6 | 1307 | 102 | ||
| BC068116.1 | - | 6 - 7 | 545 | 6809443 | 102 | |
| BC098097.1 | - | 6 - 7 | 559 | 6403012 | 102 | |
Set Membership: |
|