Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01172074_s1
          See other OLFR934 GE Assays ›
          Gene Symbol:
          Olfr934
          Gene Name:
          olfactory receptor 934
          Gene Aliases:
          MOR224-6
          Chromosome Location:
          Chr.9: 38982110 - 38983042 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01172074_s1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr934
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146442.1 NP_666653.1 1 831 119
          GenBank mRNA BC113124.1 - 1 830 119

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr934

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P527697 NM_146442.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P527697 BC113124.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P527697 NM_146442.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P527697 BC113124.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P135015 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P527697 NM_146442.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P527697 BC113124.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P527697 NM_146442.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P527697 BC113124.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P527697 NM_146442.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P527697 BC113124.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P527697 NM_146442.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P527697 BC113124.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26436876 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26436892 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26436890 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26436868 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26436880 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26436889 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26436886 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26436873 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26436883 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26436853 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26436852 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26436867 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26436888 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26436877 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26436864 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26436879 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26436854 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26436857 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26436856 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26436858 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26436893 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26436874 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26436849 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26436851 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26436870 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26436859 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26436866 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26436875 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26436872 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26436881 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P135015 - -
          Whole Mouse Genome G4122A A_52_P527697 NM_146442.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P527697 BC113124.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P527697 NM_146442.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P527697 BC113124.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P139051 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P135015 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07725124 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07545044 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07545045 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07725135 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07725128 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436828 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436856 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436875 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436853 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436831 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436814 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436881 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436835 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436889 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436873 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436808 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436806 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436846 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436838 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436811 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436899 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436844 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436827 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436833 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436879 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436817 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436880 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436797 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436890 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436792 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436798 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436849 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436805 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436893 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436864 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436791 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436858 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436852 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436800 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436810 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436854 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436789 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436795 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436866 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436876 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436801 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436794 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436850 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436851 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436843 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436892 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436868 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436874 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436895 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436836 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436837 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436886 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26436881 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26436874 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26436856 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26436880 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26436858 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26436890 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26436884 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26436852 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26436873 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26436895 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26436875 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26436898 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07627030 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26436867 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26436889 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26436864 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26436853 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26436843 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26436851 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26436876 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26436886 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26436868 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26436859 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26436901 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26436849 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07640397 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26436854 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26436900 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26436863 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26436877 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26436892 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26436850 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26436844 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26436893 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26436872 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26436899 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26436846 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10592383 - -
          Affymetrix Mouse Gene 1.1 ST Array 10592382 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 645154 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 4200110 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004200110 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1219001 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1219001 - -
          mouse-6 v1.1 (Illumina) scl36014.1.1_209-S - -
          Illumina Sentrix mouse-6.v1 bead array 4200110 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl36014.1.1_209-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4200110 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1219001 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1219001 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline