Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01172741_g1
          See other PLDI GE Assays ›
          Gene Symbol:
          Pldi
          Gene Name:
          polymorphic derived intron containing
          Gene Aliases:
          1700125F08Rik, Poldi
          Chromosome Location:
          Chr.10: 60928226 - 60938132 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01172741_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pldi
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_033616.1 2 - 3 273 69
          GenBank mRNA AK007277.1 - 2 - 3 276 69

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the AK007277.1 transcripts.

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Mus musculus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          sperm motility
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P449638 NR_033616.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P449638 AK007277.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P449638 NR_033616.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P449638 AK007277.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P111961 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P111961 - -
          Whole Mouse Genome G4122A A_51_P449638 NR_033616.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P449638 AK007277.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P449638 NR_033616.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P449638 AK007277.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P100411 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P111961 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27177653 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27177571 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08357826 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27177683 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27177651 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27177543 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27177662 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27177556 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08299326 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27177589 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27177575 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27177549 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27177677 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27177687 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27177563 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08821150 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27177533 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27177577 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27177544 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27177567 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08821148 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27177592 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27177668 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08957493 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27177558 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27177686 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27177654 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27177548 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27177593 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27177672 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27177551 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08821136 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08821143 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27177670 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08635730 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08821147 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27177531 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27177665 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27177596 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08450742 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08281015 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27177688 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08357831 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27177597 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27177594 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08392907 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27177658 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27177659 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27177555 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08957486 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08357847 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27177530 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08821135 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27177570 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27177590 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27177674 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27177564 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08635733 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27177581 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08821139 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27177598 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27177559 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1432460_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10369409 - -
          Mouse Genome 430 2.0 Array 1432460_at - -
          Mouse Genome 430 2.0 Array 1432460_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1000015734.mm.1 NR_033616.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000015734.mm.1 NR_033616.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000015734.mm.1 TC1000002237.mm.1 MTA TC
          Mouse Expression Set 430 1432460_at - -
          Mouse Expression Set 430 1432460_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1432460_at - -
          Affymetrix HT Mouse Genome 430B Array 1432460_PM_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline