Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Cic | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AB093224.1 | - | 2 - 3 | 444 | 68 | |
AF363690.1 | - | 7 - 8 | 1174 | 68 | ||
AK147276.1 | - | 7 - 8 | 1807 | 68 | ||
AK147535.1 | - | 8 - 9 | 3925 | 68 | ||
BC058665.1 | - | 4 - 5 | 692 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |