Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01174111_s1
          See other OLFR836 GE Assays ›
          Gene Symbol:
          Olfr836
          Gene Name:
          olfactory receptor 836
          Gene Aliases:
          MOR152-3
          Chromosome Location:
          Chr.9: 19120957 - 19121911 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01174111_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr836
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146564.2 NP_666775.1 1 900 130
          GenBank mRNA BC106988.1 - 1 900 40043348 130
          BC106989.1 - 1 900 40043350 130

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr836

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P402443 BC106988.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P402443 NM_146564.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P402443 BC106989.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P402443 BC106988.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P402443 NM_146564.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P402443 BC106989.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P101030 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P402443 BC106988.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P402443 NM_146564.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P402443 BC106989.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P402443 BC106988.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P402443 NM_146564.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P402443 BC106989.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P402443 BC106988.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P402443 NM_146564.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P402443 BC106989.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P402443 BC106988.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P402443 NM_146564.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P402443 BC106989.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26339218 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26339207 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26339210 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26339212 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26339237 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26339220 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26339216 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26339243 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26339240 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26339234 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26339244 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26339214 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26339217 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26339238 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26339213 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26339224 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26339235 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26339226 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26339209 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26339242 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P101030 - -
          Whole Mouse Genome G4122A A_51_P402443 BC106988.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P402443 NM_146564.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P402443 BC106989.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P402443 BC106988.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P402443 NM_146564.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P402443 BC106989.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P106404 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P113543 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P101030 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07930043 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07930050 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07930033 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07930047 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07930045 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07930040 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26339238 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26339245 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26339209 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26339252 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26339224 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26339247 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26339253 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26339249 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26339220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26339250 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26339255 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26339259 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26339256 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26339243 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26339265 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26339216 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26339261 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26339242 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26339213 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26339257 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26339237 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26339226 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26339240 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26339263 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26339217 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26339210 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08012913 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26339247 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26339207 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26339210 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26339213 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26339218 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26339250 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26339261 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26339224 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26339257 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26339263 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07930029 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08012916 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26339242 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26339245 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26339226 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26339249 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08105026 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26339222 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26339255 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26339252 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26339265 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26339237 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26339259 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26339238 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26339266 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26339209 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26339253 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26339256 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26339208 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26339217 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08165665 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26339220 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26339216 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26339243 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10583389 - -
          Affymetrix Mouse Gene 1.1 ST Array 10583388 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 415566 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 7000471 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0007000471 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1240260 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1240260 - -
          mouse-6 v1.1 (Illumina) scl7637.1.1_330-S - -
          Illumina Sentrix mouse-6.v1 bead array 7000471 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl7637.1.1_330-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 7000471 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1240260 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1240260 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline