Search
Search
| Gene Symbol: Ubl7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122873.1 | NP_001116345.1 | 7 - 8 | 766 | 57 | |
| NM_027086.3 | NP_081362.2 | 7 - 8 | 714 | 57 | ||
| GenBank mRNA | AK154496.1 | - | 7 - 8 | 724 | 57 | |
| AK179100.1 | - | 3 - 4 | 278 | 57 | ||
| AK183337.1 | - | 3 - 4 | 278 | 57 | ||
| AK207226.1 | - | 3 - 4 | 278 | 57 | ||
| AK209878.1 | - | 3 - 4 | 298 | 57 | ||
| AK212304.1 | - | 3 - 4 | 279 | 57 | ||
| AK214469.1 | - | 1 - 2 | 126 | 57 | ||
| BC016456.1 | - | 6 - 7 | 700 | 4912715 | 57 | |
| BC094047.1 | - | 6 - 7 | 703 | 5253101 | 57 | |
| EL608156.1 | - | 4 - 5 | 57 | |||
Set Membership: |
|