Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01175474_g1
          See other CATSPERG2 GE Assays ›
          Gene Symbol:
          Catsperg2
          Gene Name:
          catsper channel auxiliary subunit gamma 2
          Gene Aliases:
          1700067C01Rik, CATSPERG
          Chromosome Location:
          Chr.7: 29697217 - 29727048 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01175474_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Catsperg2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_029714.3 NP_083990.2 14 - 15 1694 71
          GenBank mRNA AK077059.1 - 14 - 15 1676 71
          AK138347.1 - 10 - 11 1200 71
          BC144969.1 - 14 - 15 1674 71
          GQ225581.1 - 13 - 14 1595 71

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Catsperg2

          Panther Classification:

          Molecular Function -

          ion channel transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          multicellular organism development
          spermatogenesis
          biological_process
          cell differentiation
          protein binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0700024159.mm.1 NM_029714.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024159.mm.1 NM_029714.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024159.mm.1 TC0700002727.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700024165.mm.1 NM_029714.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024165.mm.1 NM_029714.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024165.mm.1 TC0700002727.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700024172.mm.1 NM_029714.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024172.mm.1 NM_029714.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024172.mm.1 TC0700002727.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700024173.mm.1 NM_029714.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024173.mm.1 NM_029714.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024173.mm.1 TC0700002727.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700024163.mm.1 NM_029714.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024163.mm.1 NM_029714.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024163.mm.1 TC0700002727.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700024164.mm.1 NM_029714.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024164.mm.1 NM_029714.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024164.mm.1 TC0700002727.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700024168.mm.1 NM_029714.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024168.mm.1 NM_029714.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024168.mm.1 TC0700002727.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700024169.mm.1 NM_029714.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024169.mm.1 NM_029714.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024169.mm.1 TC0700002727.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700024171.mm.1 NM_029714.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024171.mm.1 NM_029714.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024171.mm.1 TC0700002727.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700024174.mm.1 NM_029714.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024174.mm.1 NM_029714.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024174.mm.1 TC0700002727.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700024158.mm.1 NM_029714.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024158.mm.1 NM_029714.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024158.mm.1 TC0700002727.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700024161.mm.1 NM_029714.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024161.mm.1 NM_029714.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024161.mm.1 TC0700002727.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700024167.mm.1 NM_029714.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024167.mm.1 NM_029714.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024167.mm.1 TC0700002727.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700024160.mm.1 NM_029714.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024160.mm.1 NM_029714.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024160.mm.1 TC0700002727.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700024162.mm.1 NM_029714.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024162.mm.1 NM_029714.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024162.mm.1 TC0700002727.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700024166.mm.1 NM_029714.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024166.mm.1 NM_029714.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024166.mm.1 TC0700002727.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline