Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01175646_g1
          See other CATSPERG1 GE Assays ›
          Gene Symbol:
          Catsperg1
          Gene Name:
          catsper channel auxiliary subunit gamma 1
          Gene Aliases:
          A230107C01Rik, Catsperg
          Chromosome Location:
          Chr.7: 29181532 - 29215631 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01175646_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Catsperg1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001164658.1 NP_001158130.1 19 - 20 2428 75
          GenBank mRNA AK039202.1 - 11 - 12 1959 75
          AK133307.1 - 19 - 20 2209 75
          BC172039.1 - 13 - 14 1468 75

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700002717.mm.1

          Set Membership:

          Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Probe spans exons

          Panther Classification:

          Molecular Function -

          ion channel transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0700024007.mm.1 NM_001164658.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024007.mm.1 NM_001164658.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024007.mm.1 TC0700002717.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700023989.mm.1 NM_001164658.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700023989.mm.1 NM_001164658.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700023989.mm.1 TC0700002717.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700023994.mm.1 NM_001164658.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700023994.mm.1 NM_001164658.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700023994.mm.1 TC0700002717.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700023996.mm.1 NM_001164658.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700023996.mm.1 NM_001164658.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700023996.mm.1 TC0700002717.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700024016.mm.1 NM_001164658.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024016.mm.1 NM_001164658.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024016.mm.1 TC0700002717.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700024018.mm.1 NM_001164658.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024018.mm.1 NM_001164658.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024018.mm.1 TC0700002717.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700023977.mm.1 NM_001164658.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700023977.mm.1 NM_001164658.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700023977.mm.1 TC0700002717.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700024000.mm.1 NM_001164658.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024000.mm.1 NM_001164658.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024000.mm.1 TC0700002717.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700024028.mm.1 NM_001164658.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024028.mm.1 NM_001164658.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024028.mm.1 TC0700002717.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700023980.mm.1 NM_001164658.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700023980.mm.1 NM_001164658.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700023980.mm.1 TC0700002717.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700023986.mm.1 NM_001164658.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700023986.mm.1 NM_001164658.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700023986.mm.1 TC0700002717.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700024006.mm.1 NM_001164658.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024006.mm.1 NM_001164658.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024006.mm.1 TC0700002717.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700024015.mm.1 NM_001164658.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024015.mm.1 NM_001164658.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024015.mm.1 TC0700002717.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700024002.mm.1 NM_001164658.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024002.mm.1 NM_001164658.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024002.mm.1 TC0700002717.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700023981.mm.1 NM_001164658.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700023981.mm.1 NM_001164658.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700023981.mm.1 TC0700002717.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700023990.mm.1 NM_001164658.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700023990.mm.1 NM_001164658.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700023990.mm.1 TC0700002717.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700023999.mm.1 NM_001164658.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700023999.mm.1 NM_001164658.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700023999.mm.1 TC0700002717.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700024012.mm.1 NM_001164658.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024012.mm.1 NM_001164658.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024012.mm.1 TC0700002717.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700024017.mm.1 NM_001164658.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024017.mm.1 NM_001164658.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024017.mm.1 TC0700002717.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700024019.mm.1 NM_001164658.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024019.mm.1 NM_001164658.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024019.mm.1 TC0700002717.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700024026.mm.1 NM_001164658.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024026.mm.1 NM_001164658.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024026.mm.1 TC0700002717.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700024029.mm.1 NM_001164658.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024029.mm.1 NM_001164658.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024029.mm.1 TC0700002717.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700024032.mm.1 NM_001164658.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024032.mm.1 NM_001164658.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024032.mm.1 TC0700002717.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700024010.mm.1 NM_001164658.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024010.mm.1 NM_001164658.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024010.mm.1 TC0700002717.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700024020.mm.1 NM_001164658.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024020.mm.1 NM_001164658.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024020.mm.1 TC0700002717.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700024021.mm.1 NM_001164658.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024021.mm.1 NM_001164658.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024021.mm.1 TC0700002717.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700024023.mm.1 NM_001164658.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024023.mm.1 NM_001164658.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024023.mm.1 TC0700002717.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700023998.mm.1 NM_001164658.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700023998.mm.1 NM_001164658.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700023998.mm.1 TC0700002717.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700024005.mm.1 NM_001164658.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024005.mm.1 NM_001164658.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024005.mm.1 TC0700002717.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700024014.mm.1 NM_001164658.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024014.mm.1 NM_001164658.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024014.mm.1 TC0700002717.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700024025.mm.1 NM_001164658.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024025.mm.1 NM_001164658.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024025.mm.1 TC0700002717.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline