| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0900019107.mm.1 |
NR_033560.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900019107.mm.1 |
NR_033560.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900019107.mm.1 |
TC0900002290.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900019105.mm.1 |
NR_033560.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900019105.mm.1 |
NR_033560.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900019105.mm.1 |
TC0900002290.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900019110.mm.1 |
NR_033560.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900019110.mm.1 |
NR_033560.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900019110.mm.1 |
TC0900002290.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900019108.mm.1 |
NR_033560.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900019108.mm.1 |
NR_033560.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900019108.mm.1 |
TC0900002290.mm.1 |
MTA TC |