Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01176349_g1
          See other VWA5A GE Assays ›
          Gene Symbol:
          Vwa5a
          Gene Name:
          von Willebrand factor A domain containing 5A
          Gene Aliases:
          5830475I06Rik, AW552491, BCSC-1, E130119J22, Loh11cr2a
          Chromosome Location:
          Chr.9: 38718268 - 38743337 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01176349_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Vwa5a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001145957.1 NP_001139429.1 3 - 4 386 61
          NM_172767.3 NP_766355.3 4 - 5 462 61
          GenBank mRNA AK019914.1 - 4 - 5 416 61
          AK041397.1 - 4 - 5 429 61
          AK053665.1 - 4 - 5 406 61
          AK054497.1 - 4 - 5 435 61
          AK136934.1 - 4 - 5 435 61
          AK161193.1 - 3 - 4 373 61
          AY366502.1 - 4 - 5 432 61
          BC065153.1 - 3 - 4 316 6418714 61

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900000412.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Vwa5a

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0900002820.mm.1 NM_001145957.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900002820.mm.1 NM_001145957.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900002820.mm.1 NM_172767.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900002820.mm.1 NM_172767.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900002820.mm.1 TC0900000412.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900002822.mm.1 NM_172767.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900002822.mm.1 NM_172767.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900002822.mm.1 TC0900000412.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900002819.mm.1 NM_001145957.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900002819.mm.1 NM_001145957.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900002819.mm.1 NM_172767.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900002819.mm.1 NM_172767.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900002819.mm.1 TC0900000412.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900002825.mm.1 NM_001145957.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900002825.mm.1 NM_001145957.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900002825.mm.1 NM_172767.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900002825.mm.1 NM_172767.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900002825.mm.1 TC0900000412.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900002826.mm.1 NM_001145957.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900002826.mm.1 NM_001145957.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900002826.mm.1 NM_172767.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900002826.mm.1 NM_172767.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900002826.mm.1 TC0900000412.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900002823.mm.1 NM_001145957.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900002823.mm.1 NM_001145957.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900002823.mm.1 NM_172767.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900002823.mm.1 NM_172767.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900002823.mm.1 TC0900000412.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900002824.mm.1 NM_001145957.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900002824.mm.1 NM_001145957.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900002824.mm.1 NM_172767.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900002824.mm.1 NM_172767.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900002824.mm.1 TC0900000412.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 910497 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline