Search
Search

Psmc4| Gene Symbol: Psmc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011874.2 | NP_036004.2 | 9 - 10 | 1168 | 110 | |
| GenBank mRNA | AK050677.1 | - | 9 - 10 | 1146 | 110 | |
| AK077507.1 | - | 9 - 10 | 1131 | 110 | ||
| AK153991.1 | - | 9 - 10 | 1151 | 110 | ||
| AK160644.1 | - | 9 - 10 | 1130 | 110 | ||
| AK160718.1 | - | 9 - 10 | 1122 | 110 | ||
| AK167041.1 | - | 9 - 10 | 1113 | 110 | ||
| AK167528.1 | - | 9 - 10 | 1123 | 110 | ||
| AK169224.1 | - | 9 - 10 | 1131 | 110 | ||
| AK169244.1 | - | 9 - 10 | 1630 | 110 | ||
| BC012411.1 | - | 1 - 2 | 390 | 110 | ||
| BC012708.1 | - | 9 - 10 | 1129 | 4211677 | 110 | |
| BC092265.1 | - | 9 - 10 | 1112 | 3671250 | 110 | |
| BC094063.1 | - | 4 - 5 | 486 | 110 | ||
Set Membership: |
|