Search
Search

Tmprss5| Gene Symbol: Tmprss5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030709.2 | NP_109634.2 | 4 - 5 | 410 | 100 | |
| GenBank mRNA | AK029714.1 | - | 5 - 6 | 1257 | 100 | |
| BC132151.1 | - | 3 - 4 | 356 | 40130428 | 100 | |
| BC145027.1 | - | 3 - 4 | 338 | 9053575 | 100 | |
Set Membership: |
|