Search
Search

1810046K07Rik| Gene Symbol: 1810046K07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027217.1 | NP_081493.1 | 3 - 4 | 260 | 60 | |
| GenBank mRNA | AK007796.1 | - | 3 - 4 | 263 | 60 | |
| BC120857.1 | - | 1 - 2 | 44 | 40129780 | 60 | |
| BC120859.1 | - | 1 - 2 | 44 | 40129782 | 60 | |
Set Membership: |
|