Search
Search

Specc1l| Gene Symbol: Specc1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145826.1 | NP_001139298.1 | 17 - 18 | 3617 | 80 | |
| NM_153406.3 | NP_700455.3 | 14 - 15 | 3388 | 80 | ||
| GenBank mRNA | AB093236.1 | - | 17 - 18 | 3617 | 80 | |
| AK015541.1 | - | 3 - 4 | 353 | 80 | ||
| AK016555.1 | - | 3 - 4 | 354 | 80 | ||
| AY884297.1 | - | 14 - 15 | 3389 | 80 | ||
| BC037464.1 | - | 11 - 12 | 2475 | 4194868 | 80 | |
| BC072595.1 | - | 13 - 14 | 3227 | 6408832 | 80 | |
Set Membership: |
|