Search
Search

Zfp526| Gene Symbol: Zfp526 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175436.4 | NP_780645.1 | 1 - 2 | 52 | 73 | |
| GenBank mRNA | AK050839.1 | - | 1 - 2 | 53 | 73 | |
| AK051798.1 | - | 1 - 2 | 48 | 73 | ||
Set Membership: |
|