Search
Search

Rpusd4| Gene Symbol: Rpusd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028040.2 | NP_082316.1 | 5 - 6 | 802 | 67 | |
| GenBank mRNA | AK010322.1 | - | 5 - 6 | 819 | 67 | |
| AK156574.1 | - | 5 - 6 | 819 | 67 | ||
| BC061256.1 | - | 5 - 6 | 803 | 30251586 | 67 | |
Set Membership: |
|