Search
Search
| Gene Symbol: Snx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019727.2 | NP_062701.2 | 5 - 6 | 627 | 105 | |
| GenBank mRNA | AB019214.1 | - | 5 - 6 | 567 | 105 | |
| AK149850.1 | - | 5 - 6 | 539 | 105 | ||
| AK154079.1 | - | 5 - 6 | 540 | 105 | ||
| AK155746.1 | - | 5 - 6 | 515 | 105 | ||
| AK161241.1 | - | 5 - 6 | 608 | 105 | ||
| AK167177.1 | - | 5 - 6 | 582 | 105 | ||
| AK168742.1 | - | 5 - 6 | 620 | 105 | ||
| AK219007.1 | - | 3 - 4 | 157 | 105 | ||
| BC066044.1 | - | 5 - 6 | 584 | 6807363 | 105 | |
| BC066189.1 | - | 5 - 6 | 518 | 30471747 | 105 | |
Set Membership: |
|