Search
Search

Nr2e3| Gene Symbol: Nr2e3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013708.4 | NP_038736.1 | 5 - 6 | 900 | 63 | |
| GenBank mRNA | AF148129.1 | - | 5 - 6 | 762 | 63 | |
| AF204053.1 | - | 5 - 6 | 900 | 63 | ||
| AK044718.1 | - | 4 - 5 | 1032 | 63 | ||
| AK045788.1 | - | 3 - 4 | 643 | 63 | ||
| AK045859.1 | - | 3 - 4 | 643 | 63 | ||
| AK053560.1 | - | 5 - 6 | 868 | 63 | ||
| BC017521.1 | - | 5 - 6 | 894 | 4512088 | 63 | |
| BC047150.1 | - | 2 - 3 | 1587 | 63 | ||
| BC057958.1 | - | 2 - 3 | 1627 | 63 | ||
Set Membership: |
|