Search
Search

Pias1| Gene Symbol: Pias1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019663.3 | NP_062637.2 | 7 - 8 | 932 | 99 | |
| GenBank mRNA | AF077950.1 | - | 7 - 8 | 924 | 99 | |
| AK013753.1 | - | 3 - 4 | 267 | 99 | ||
| AK075708.1 | - | 7 - 8 | 933 | 99 | ||
| AK153871.1 | - | 7 - 8 | 937 | 99 | ||
| BC051417.1 | - | 7 - 8 | 929 | 6515027 | 99 | |
Set Membership: |
|