Search
Search

Vps26b| Gene Symbol: Vps26b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178027.4 | NP_821170.1 | 3 - 4 | 1016 | 95 | |
| GenBank mRNA | AK010160.1 | - | 3 - 4 | 950 | 95 | |
| AK031589.1 | - | 3 - 4 | 1019 | 95 | ||
| AK150929.1 | - | 3 - 4 | 937 | 95 | ||
| AK155578.1 | - | 3 - 4 | 977 | 95 | ||
| AK165539.1 | - | 3 - 4 | 955 | 95 | ||
| AK171154.1 | - | 3 - 4 | 975 | 95 | ||
| AK215961.1 | - | 1 - 2 | 61 | 95 | ||
| BC029758.1 | - | 1 - 2 | 109 | 95 | ||
| BC049180.1 | - | 3 - 4 | 979 | 6314347 | 95 | |
| BC052721.1 | - | 3 - 4 | 940 | 6809853 | 95 | |
| BC062972.1 | - | 3 - 4 | 940 | 6809878 | 95 | |
Set Membership: |
|