Search
Search

Matn1| Gene Symbol: Matn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010769.2 | NP_034899.2 | 6 - 7 | 1406 | 78 | |
| GenBank mRNA | AK014342.1 | - | 6 - 7 | 1406 | 78 | |
| BC047140.1 | - | 6 - 7 | 1373 | 6315509 | 78 | |
| U35035.1 | - | 6 - 7 | 1367 | 78 | ||
Set Membership: |
|