Search
Search
| Gene Symbol: Ogdh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252282.1 | NP_001239211.1 | 17 - 18 | 2342 | 79 | |
| NM_001252283.1 | NP_001239212.1 | 17 - 18 | 2330 | 79 | ||
| NM_001252287.1 | NP_001239216.1 | 16 - 17 | 2373 | 79 | ||
| NM_001252288.1 | NP_001239217.1 | 16 - 17 | 2345 | 79 | ||
| NM_010956.4 | NP_035086.2 | 16 - 17 | 2297 | 79 | ||
| GenBank mRNA | AK147289.1 | - | 16 - 17 | 2298 | 79 | |
| AK150009.1 | - | 16 - 17 | 2263 | 79 | ||
| AK169286.1 | - | 16 - 17 | 2289 | 79 | ||
| AK220536.1 | - | 17 - 18 | 79 | |||
| BC013670.1 | - | 16 - 17 | 2231 | 4188988 | 79 | |
| BC025040.1 | - | 1 - 2 | 97 | 79 | ||
| BC029143.1 | - | 1 - 2 | 107 | 79 | ||
| BC031165.1 | - | 1 - 2 | 76 | 79 | ||
| BC049104.1 | - | 16 - 17 | 2298 | 6417392 | 79 | |
| BC057354.1 | - | 17 - 18 | 2258 | 6827509 | 79 | |
Set Membership: |
|