Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01180869_s1
          See other SFN GE Assays ›
          Gene Symbol:
          Sfn
          Gene Name:
          stratifin
          Gene Aliases:
          Er, Mme1, Ywhas
          Chromosome Location:
          Chr.4: 133600556 - 133602168 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01180869_s1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sfn
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_018754.2 NP_061224.2 1 332 71
          GenBank mRNA CX224942.1 - 1 395 71

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
          This assay is in the 5'UTR region of the NM_018754.2 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Sfn

          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of cyclin-dependent protein serine/threonine kinase activity
          release of cytochrome c from mitochondria
          keratinocyte development
          intrinsic apoptotic signaling pathway in response to DNA damage
          regulation of epidermal cell division
          negative regulation of keratinocyte proliferation
          keratinocyte differentiation
          positive regulation of cell growth
          keratinization
          negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
          skin development
          positive regulation of epidermal cell differentiation
          positive regulation of protein export from nucleus
          regulation of cell cycle
          establishment of skin barrier
          protein binding
          protein kinase binding
          protein domain specific binding
          phosphoprotein binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23318412 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23318415 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23318414 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23318413 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2170514 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 226675 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2170514 - -
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P334191 AF058798.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P334191 AF152893.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P334191 AK146490.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P334191 AK169358.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P334191 NM_018754.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P213940 AF058798.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P334188 AF058798.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P334188 AF152893.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P334188 AK146490.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P334188 AK169358.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P334188 NM_018754.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23318432 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23318459 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23318418 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23318419 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23318448 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23318465 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23318434 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23318439 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23318455 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23318464 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23318436 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23318447 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23318468 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23318470 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23318435 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23318452 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23318414 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23318458 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23318415 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23318450 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23318431 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23318462 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23318442 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23318440 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23318441 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23318444 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23318445 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23318412 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23318463 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23318456 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23318453 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23318417 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23318438 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23318437 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23318454 - -
          Whole Mouse Genome G4122A A_52_P334188 AF058798.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P334188 AF152893.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P334188 AK146490.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P334188 AK169358.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P334188 NM_018754.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P213940 AF058798.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P334191 AF058798.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P334191 AF152893.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P334191 AK146490.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P334191 AK169358.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P334191 NM_018754.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P110442 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23318413 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23318415 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23318412 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23318414 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318434 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318445 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318368 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318439 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318441 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318465 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318371 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318365 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318417 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318456 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318400 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318419 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318407 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318370 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318415 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318362 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318432 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318412 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318458 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318409 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318394 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318373 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318463 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318405 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318453 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318396 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318435 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318464 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318462 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318450 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318459 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318449 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318440 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318395 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318414 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318392 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318363 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318437 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318436 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318431 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318444 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03371816 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23318417 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23318431 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23318411 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23318434 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23318409 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23318394 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04091405 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23318437 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03459836 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23318435 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03969353 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23318396 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03205832 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23318464 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23318465 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23318448 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23318456 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23318445 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23318392 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23318400 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23318432 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23318407 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23318436 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23318403 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23318412 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23318419 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23318462 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23318449 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23318408 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23318418 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23318440 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23318395 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23318458 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23318453 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23318463 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23318439 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03341920 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10572636 - -
          Affymetrix Mouse Gene 1.0 ST Array 10517069 - -
          Affymetrix Mouse Gene 1.0 ST Array 10517068 - -
          Affymetrix HT MG-430 PM Array Plate 1448612_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10572635 - -
          Affymetrix Mouse Gene 1.1 ST Array 10517067 - -
          Mouse Genome 430 2.0 Array 1448612_at - -
          Mouse Genome 430 2.0 Array 1448612_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0400030375.mm.1 TC0400003707.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400030375.mm.1 NM_018754.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400030375.mm.1 NM_018754.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400030376.mm.1 TC0400003707.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400030376.mm.1 NM_018754.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400030376.mm.1 NM_018754.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400030374.mm.1 TC0400003707.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400030374.mm.1 NM_018754.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400030374.mm.1 NM_018754.2 Refseq
          Mouse Expression Set 430 1448612_PM_at - -
          Mouse Expression Set 430 1448612_at - -
          Murine Genome U74v2 Set 96704_PM_at - -
          Murine Genome U74v2 Set 96704_at - -
          Mouse Genome 430A 2.0 Array 1448612_PM_at - -
          Mouse Genome 430A 2.0 Array 1448612_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6290301 - -
          Illumina mouse-6 v1.0 expression beadchip 3520397 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006290301 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003520397 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2648168 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2763282 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2763282 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2648168 - -
          mouse-6 v1.1 (Illumina) scl0055948.1_66-S - -
          mouse-6 v1.1 (Illumina) scl055948.2_144-S - -
          Illumina Sentrix mouse-6.v1 bead array 3520397 - -
          Illumina Sentrix mouse-6.v1 bead array 6290301 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl055948.2_144-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0055948.1_66-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3520397 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6290301 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2867084 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2648169 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2648169 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2867084 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-d4j86:80/100.66.77.21:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline