Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zmiz2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005867.1 | NP_001005867.1 | 13 - 14 | 1989 | 111 | |
NM_028601.3 | NP_082877.2 | 13 - 14 | 2085 | 111 | ||
GenBank mRNA | BC024081.1 | - | 12 - 13 | 1835 | 111 | |
BC040745.1 | - | 4 - 5 | 544 | 111 | ||
BC060652.1 | - | 13 - 14 | 1927 | 30354393 | 111 | |
BC063069.1 | - | 5 - 6 | 690 | 111 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |