Search
Search

Il5ra| Gene Symbol: Il5ra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008370.2 | NP_032396.1 | 7 - 8 | 814 | 77 | |
| GenBank mRNA | BC125374.1 | - | 6 - 7 | 580 | 40129889 | 77 |
| BC145043.1 | - | 6 - 7 | 580 | 9053591 | 77 | |
| D90205.1 | - | 7 - 8 | 814 | 77 | ||
Set Membership: |
|