Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Gene Symbol: Slc41a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037493.2 | NP_001032570.1 | 2 - 3 | 511 | 79 | |
NM_027868.2 | NP_082144.2 | 3 - 4 | 543 | 79 | ||
GenBank mRNA | AK003140.1 | - | 2 - 3 | 511 | 79 | |
BC011108.1 | - | 3 - 4 | 459 | 4011836 | 79 | |
CN699671.1 | - | 3 - 4 | 457 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |