Search
Search
| Gene Symbol: Gck | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001287386.1 | NP_001274315.1 | 3 - 4 | 485 | 64 | |
| NM_010292.5 | NP_034422.2 | 3 - 4 | 843 | 64 | ||
| GenBank mRNA | AK142698.1 | - | 3 - 4 | 484 | 64 | |
| BC011139.1 | - | 3 - 4 | 474 | 4195943 | 64 | |
| L38990.1 | - | 3 - 4 | 361 | 64 | ||
Set Membership: |
|