| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0600010739.mm.1 |
NM_008359.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600010739.mm.1 |
NM_008359.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600010739.mm.1 |
TC0600001347.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600010740.mm.1 |
NM_008359.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600010740.mm.1 |
NM_008359.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600010740.mm.1 |
TC0600001347.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600010741.mm.1 |
NM_008359.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600010741.mm.1 |
NM_008359.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600010741.mm.1 |
TC0600001347.mm.1 |
MTA TC |