Search
Search

Anxa4| Gene Symbol: Anxa4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013471.2 | NP_038499.2 | 4 - 5 | 301 | 69 | |
| GenBank mRNA | AK032703.1 | - | 3 - 4 | 329 | 69 | |
| AK132293.1 | - | 5 - 6 | 343 | 69 | ||
| AK150486.1 | - | 4 - 5 | 262 | 69 | ||
| AK150614.1 | - | 4 - 5 | 248 | 69 | ||
| AK151054.1 | - | 4 - 5 | 248 | 69 | ||
| AK151236.1 | - | 4 - 5 | 280 | 69 | ||
| AK167338.1 | - | 4 - 5 | 302 | 69 | ||
| AK168390.1 | - | 4 - 5 | 273 | 69 | ||
| AK168487.1 | - | 4 - 5 | 282 | 69 | ||
| AK168917.1 | - | 3 - 4 | 306 | 69 | ||
| AK170447.1 | - | 4 - 5 | 247 | 69 | ||
| BC055871.1 | - | 4 - 5 | 261 | 4947415 | 69 | |
| U72941.1 | - | 4 - 5 | 272 | 69 | ||
| U95371.1 | - | 4 - 5 | 272 | 69 | ||
Set Membership: |
|