Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01184748_g1
          See other RPUSD3 GE Assays ›
          Gene Symbol:
          Rpusd3
          Gene Name:
          RNA pseudouridylate synthase domain containing 3
          Gene Aliases:
          AI527266
          Chromosome Location:
          Chr.6: 113415319 - 113419459 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01184748_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rpusd3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001033204.1 NP_001028376.1 8 - 9 861 56
          GenBank mRNA AK133871.1 - 8 - 9 828 56
          BC038081.1 - 8 - 9 802 56
          BC116825.1 - 8 - 9 861 40126144 56
          BC116827.1 - 8 - 9 861 40126146 56
          BC144733.1 - 8 - 9 847 9053281 56
          BC144734.1 - 7 - 8 939 56

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600002919.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Panther Classification:

          Molecular Function -

          RNA processing factor

          Gene Ontology Categories:

          Function(s) Process(es)

          pseudouridine synthesis
          RNA modification
          RNA binding
          pseudouridine synthase activity
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2166421 BC144733.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2166421 NM_001033204.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2166421 AK133871.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2166421 BC038081.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2157610 NM_001033204.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2157610 AK133871.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2157610 BC038081.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 51455 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 924105 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P175926 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2157610 NM_001033204.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2157610 AK133871.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2157610 BC038081.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2166421 BC144733.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2166421 NM_001033204.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2166421 AK133871.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2166421 BC038081.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P175926 - -
          Whole Mouse Genome G4122A A_52_P411507 AK076749.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P175926 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10546921 - -
          Affymetrix Mouse Gene 1.0 ST Array 10546922 - -
          Affymetrix Mouse Gene 1.0 ST Array 10546924 - -
          Affymetrix Mouse Gene 1.0 ST Array 10546927 - -
          Affymetrix Mouse Gene 1.0 ST Array 10546926 - -
          Affymetrix Mouse Gene 1.0 ST Array 10546920 - -
          Affymetrix Mouse Gene 1.0 ST Array 10546928 - -
          Affymetrix Mouse Gene 1.0 ST Array 10546923 - -
          Affymetrix Mouse Gene 1.0 ST Array 10546925 - -
          Affymetrix HT MG-430 PM Array Plate 1434626_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10546919 - -
          Mouse Genome 430 2.0 Array 1434626_PM_at - -
          Mouse Genome 430 2.0 Array 1434626_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 101122_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 101122_at - -
          Affymetrix Mouse Clariom S Array PSR0600022414.mm.1 NM_001033204.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600022414.mm.1 NM_001033204.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600022414.mm.1 TC0600002919.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600022412.mm.1 NM_001033204.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600022412.mm.1 NM_001033204.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600022412.mm.1 TC0600002919.mm.1 MTA TC
          Mouse Expression Set 430 1434626_PM_at - -
          Mouse Expression Set 430 1434626_at - -
          Murine Genome U74v2 Set 169396_PM_r_at - -
          Murine Genome U74v2 Set 169434_PM_f_at - -
          Murine Genome U74v2 Set 169434_f_at - -
          Murine Genome U74v2 Set 169396_r_at - -
          Murine Genome U74v2 Set 171525_PM_r_at - -
          Murine Genome U74v2 Set 171525_r_at - -
          Affymetrix HT Mouse Genome 430B Array 1434626_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1434626_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 752499 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1214740 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1214740 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline