Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01185019_m1
          See other PTPN6 GE Assays ›
          Gene Symbol:
          Ptpn6
          Gene Name:
          protein tyrosine phosphatase, non-receptor type 6
          Gene Aliases:
          70Z-SHP, Hcph, PTPTY-42, Ptp1C, SH-PTP1, SHP-1, hcp, me
          Chromosome Location:
          Chr.6: 124720707 - 124738709 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01185019_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ptpn6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001077705.2 NP_001071173.1 3 - 4 534 78
          NM_013545.3 NP_038573.2 3 - 4 532 78
          GenBank mRNA AK132509.1 - 3 - 4 451 78
          AK150509.1 - 3 - 4 439 78
          AK151077.1 - 3 - 4 453 78
          AK151623.1 - 3 - 4 456 78
          AK159686.1 - 3 - 4 491 78
          AK159833.1 - 3 - 4 484 78
          AK167923.1 - 3 - 4 486 78
          AK169804.1 - 3 - 4 534 78
          BC012660.1 - 3 - 4 477 4010462 78
          M68902.1 - 3 - 4 484 78
          M90389.1 - 3 - 4 434 78

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600003114.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Protein Phosphatase Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ptpn6

          Panther Classification:

          Molecular Function -

          protein phosphatase

          Gene Ontology Categories:

          Function(s) Process(es)

          hematopoietic progenitor cell differentiation
          negative regulation of humoral immune response mediated by circulating immunoglobulin
          protein dephosphorylation
          signal transduction
          cell proliferation
          positive regulation of cell proliferation
          positive regulation of phosphatidylinositol 3-kinase signaling
          dephosphorylation
          peptidyl-tyrosine phosphorylation
          cytokine-mediated signaling pathway
          cell differentiation
          platelet formation
          abortive mitotic cell cycle
          positive regulation of cell adhesion mediated by integrin
          peptidyl-tyrosine dephosphorylation
          intracellular signal transduction
          megakaryocyte development
          negative regulation of T cell proliferation
          natural killer cell mediated cytotoxicity
          negative regulation of MAP kinase activity
          negative regulation of MAPK cascade
          regulation of B cell differentiation
          negative regulation of peptidyl-tyrosine phosphorylation
          B cell receptor signaling pathway
          negative regulation of B cell receptor signaling pathway
          negative regulation of T cell receptor signaling pathway
          regulation of release of sequestered calcium ion into cytosol
          regulation of ERK1 and ERK2 cascade
          platelet aggregation
          regulation of G1/S transition of mitotic cell cycle
          phosphotyrosine binding
          phosphoprotein phosphatase activity
          protein tyrosine phosphatase activity
          transmembrane receptor protein tyrosine phosphatase activity
          protein binding
          hydrolase activity
          phosphatase activity
          SH3 domain binding
          protein kinase binding
          SH2 domain binding
          natural killer cell lectin-like receptor binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0600024096.mm.1 NM_001077705.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0600024096.mm.1 NM_013545.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0600024096.mm.1 TC0600003114.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600024095.mm.1 NM_001077705.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0600024095.mm.1 NM_013545.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0600024095.mm.1 TC0600003114.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600024097.mm.1 NM_001077705.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0600024097.mm.1 NM_013545.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0600024097.mm.1 TC0600003114.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline