Search
Search

Lpcat3| Gene Symbol: Lpcat3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145130.2 | NP_660112.1 | 7 - 8 | 898 | 75 | |
| GenBank mRNA | AB294194.1 | - | 7 - 8 | 793 | 75 | |
| AK083687.1 | - | 5 - 6 | 454 | 75 | ||
| AK098090.1 | - | 7 - 8 | 869 | 75 | ||
| AK154062.1 | - | 7 - 8 | 892 | 75 | ||
| AK165319.1 | - | 7 - 8 | 877 | 75 | ||
| AK169452.1 | - | 6 - 7 | 1015 | 75 | ||
| AY028317.1 | - | 7 - 8 | 874 | 75 | ||
| BC006753.1 | - | 7 - 8 | 826 | 3709076 | 75 | |
Set Membership: |
|